Learn More
Electron Microscopy Sciences Winlab Rack Carrier for 30 Slide Rack
SDP
Catalog No. 501927051
Encompass_Preferred
Steel wire hooks are molded to the polypropylene handle to form Slide Rack Hanger. The Slide Rack Hanger is very easy to attach or detach from the WINLAB Slide Racks.
Capacity: 30 slides
Catalog No. 50-192-7051 Supplier Electron Microscopy Sciences Supplier No. 71433DAKO
Steel wire hooks are molded to the polypropylene handle to form Slide Rack Hanger. The Slide Rack Hanger is very easy to attach or detach from the WINLAB Slide Racks.
Capacity: 30 slides