Learn More
Electron Microscopy Sciences Winlab Slide Rack LS-30, 30 Slide Capacity
SDP
Catalog No. 50931055
Encompass_Preferred
WINLAB Racks are made from Poly-ceramic that maintains its structural integrity to 200 degree C in any solvent while being microwaved. Designed for histology and cytology staining and coverslipping. They last a lot longer than any other materials and save time and money. Held rigid by the carrier in same way as Sakura/Bayer models. Preferred for Leica stainers. Handle positions: 1 vertical, 2 x 45 deg
Capacity: 30 slides
Catalog No. 50-931-055 Supplier Electron Microscopy Sciences Supplier No. 7143030
WINLAB Racks are made from Poly-ceramic that maintains its structural integrity to 200 degree C in any solvent while being microwaved. Designed for histology and cytology staining and coverslipping. They last a lot longer than any other materials and save time and money. Held rigid by the carrier in same way as Sakura/Bayer models. Preferred for Leica stainers. Handle positions: 1 vertical, 2 x 45 deg
Capacity: 30 slides