Learn More
Electron Microscopy Sciences Winlab Slide Rack LSM-30, 30 Slide Capacity
SDP
Catalog No. 50931057
Encompass_Preferred
WINLAB Racks are made from Poly-ceramic that maintains its structural integrity to 200 degree C in any solvent while being microwaved. Designed for histology and cytology staining and coverslipping. They last a lot longer than any other materials and save time and money. Held rigid by carrier and swivels. Fits Leica, VBS, Bayer, Sakura, Medite coverstaining machines and Sakura tape coverslipper
Capacity: 30 slides
Catalog No. 50-931-057 Supplier Electron Microscopy Sciences Supplier No. 7143130
WINLAB Racks are made from Poly-ceramic that maintains its structural integrity to 200 degree C in any solvent while being microwaved. Designed for histology and cytology staining and coverslipping. They last a lot longer than any other materials and save time and money. Held rigid by carrier and swivels. Fits Leica, VBS, Bayer, Sakura, Medite coverstaining machines and Sakura tape coverslipper
Capacity: 30 slides