Learn More
Med Vet International Skin Barrier Wipe 3M Cavilon No Sting 35 To 65% Strength Hexamethyldisiloxane / Isooctane / Acrylate Terpolymer / Polyphenylmethylsiloxane Individual Packet Sterile, 1.0mL, 3344, 30/Box
Catalog No. 502831305
Encompass_Preferred
3M™ Cavilon™ no sting barrier films contain a plasticizer that ensures they work around creases and bends They are alcohol-free to ensure they won't sting when applied The barrier film wipes are fast drying and non-sticky for enhanced comfort They are non-cytotoxic and hypoallergenic to be compatible with injured skin The wipes come in sterile individual packets for convenience
Catalog No. 50-283-1305 Supplier Med Vet International Supplier No. 1050201BX
3M™ Cavilon™ no sting barrier films contain a plasticizer that ensures they work around creases and bends They are alcohol-free to ensure they won't sting when applied The barrier film wipes are fast drying and non-sticky for enhanced comfort They are non-cytotoxic and hypoallergenic to be compatible with injured skin The wipes come in sterile individual packets for convenience