Learn More
Med Vet International Adhesive and Barrier Remover Adapt Wipe 50 per Box, 7760, 50/Box
Catalog No. 503045180
Encompass_Preferred
Removes from skin residue left behind by ostomy adhesives, wound care adhesives and patches that deliver pharmaceutical transdermal drugs Adapt universal remover wipes are silicone-based, alcohol-free and hypoallergenic for comfortable use Uses gentle solvent that quickly evaporates so that skin is ready for new pouching system, adhesive wound care item or patch 1½- x 2-inch wipes come in individual packets; there are 50 packets per box
Catalog No. 50-304-5180 Supplier Med Vet International Supplier No. 953690BX
Removes from skin residue left behind by ostomy adhesives, wound care adhesives and patches that deliver pharmaceutical transdermal drugs Adapt universal remover wipes are silicone-based, alcohol-free and hypoallergenic for comfortable use Uses gentle solvent that quickly evaporates so that skin is ready for new pouching system, adhesive wound care item or patch 1½- x 2-inch wipes come in individual packets; there are 50 packets per box