Learn More
Med Vet International Adhesive Remover Esenta™ Sting Free Wipe, 423391, 25/Box
Catalog No. 503044349
Encompass_Preferred
Wipes for fast and easy adhesive release Reduces painful skin stripping and skin damage Quickly removes all adhesive residue Feels silky and smooth, no residue left behind 100% silicone-based Chlorhexidine gluconate (CHG) compatible There are 25 individually wrapped wipes in a box
Catalog No. 50-304-4349 Supplier Med Vet International Supplier No. 1218925BX
Wipes for fast and easy adhesive release Reduces painful skin stripping and skin damage Quickly removes all adhesive residue Feels silky and smooth, no residue left behind 100% silicone-based Chlorhexidine gluconate (CHG) compatible There are 25 individually wrapped wipes in a box