Learn More
Med Vet International Adhesive Remover Remove™ Wipe, 403100, 1000/Case
Catalog No. 503044989
Encompass_Preferred
Aloe moisturizes and conditions skin without greasiness Gently cleans hydrocolloid, acrylic and rubber-based adhesive residues from skin without irritation, discomfort or trauma Applies easily and removes quickly with less need for reapplication Nonirritating and nonsensitizing Can be used anywhere there's a need for an adhesive dressing Cost-effective and convenient single-use wipes Reduces adhesive trauma to the skin by thoroughly dissolving dressing, tape and appliance adhesives Helps maintain proper skin integrity while removing adhesives and residues by reducing adhesive trauma
Catalog No. 50-304-4989 Supplier Med Vet International Supplier No. 211501CS
Aloe moisturizes and conditions skin without greasiness Gently cleans hydrocolloid, acrylic and rubber-based adhesive residues from skin without irritation, discomfort or trauma Applies easily and removes quickly with less need for reapplication Nonirritating and nonsensitizing Can be used anywhere there's a need for an adhesive dressing Cost-effective and convenient single-use wipes Reduces adhesive trauma to the skin by thoroughly dissolving dressing, tape and appliance adhesives Helps maintain proper skin integrity while removing adhesives and residues by reducing adhesive trauma