Learn More
Med Vet International Adhesive Remover Safe n Simple™ Wipe, SNS00550, 50/Box
Catalog No. 503045422
Encompass_Preferred
No-rinse design, so water is not needed to remove adhesive from wound care items and pharmaceutical transdermal patches Designed for easily irritated skin Wipes are 5 x 7 inches
Catalog No. 50-304-5422 Supplier Med Vet International Supplier No. 977963BX
No-rinse design, so water is not needed to remove adhesive from wound care items and pharmaceutical transdermal patches Designed for easily irritated skin Wipes are 5 x 7 inches