Learn More
Med Vet International Hand Sanitizing Wipe Purell 100 Count Ethyl Alcohol Wipe Individual Packet, 9022-10, 1/Box
Catalog No. 503045055
Encompass_Preferred
The Purell hand sanitizing wipes contain 62% ethyl alcohol which kill 99.99 percent of germs and bacteria on hands The alcohol wipes work within 15 seconds so you can get back to work fast The wipes can be used without water or towels for your convenience They come wrapped in individual packets, remaining moist until use The wipes measure 5 inches by 7 inches to clean most surfaces
Catalog No. 50-304-5055 Supplier Med Vet International Supplier No. 554273BX
The Purell hand sanitizing wipes contain 62% ethyl alcohol which kill 99.99 percent of germs and bacteria on hands The alcohol wipes work within 15 seconds so you can get back to work fast The wipes can be used without water or towels for your convenience They come wrapped in individual packets, remaining moist until use The wipes measure 5 inches by 7 inches to clean most surfaces