Learn More
Med Vet International Hygea Unscented Saline Wipe, Individual Packet, 6in x 4in, C22370, 24/Box
Catalog No. 503044390
Encompass_Preferred
Saline Wipe Hygea Individual Packet Saline Unscented 24 Count Recommended uses include: cleansing of eye area, cleansing of skin after post-operative care, umbilical cord area care, general skin cleansing such as minor cuts and scrapes and skin prone to diaper rash Mild, gentle cleansing that is non-irritating and suitable for sensitive skin One step easy to use application that delivers a consistent 3 mL dose of sterile saline Helps reduce the risk of microbial cross-contamination
Catalog No. 50-304-4390 Supplier Med Vet International Supplier No. 144086BX
Saline Wipe Hygea Individual Packet Saline Unscented 24 Count Recommended uses include: cleansing of eye area, cleansing of skin after post-operative care, umbilical cord area care, general skin cleansing such as minor cuts and scrapes and skin prone to diaper rash Mild, gentle cleansing that is non-irritating and suitable for sensitive skin One step easy to use application that delivers a consistent 3 mL dose of sterile saline Helps reduce the risk of microbial cross-contamination