Learn More
Benchmark Products Inc Benchmark Products Suite Wipes, Non-Sterile, 12x12 in
SDP
Catalog No. NC0168929
Encompass_Preferred
Introducing the Benchmark Products Suite Wipes, a high-performance cleaning solution designed for critical applications. These non-sterile, 12 in x 12 in, snag-resistant wipes deliver outstanding absorbance, effectively removing over 98% of surface contamination while minimizing cross-contamination potential to less than 0.02%. Crafted from advanced synthetic Ultra-Fibers, they ensure stringent cleanliness standards are upheld, making them ideal for scientific and controlled environments.
This item is not returnable. View return policy
Catalog No. NC0168929 Supplier Benchmark Products Inc Supplier No. WTTC1212C
Introducing the Benchmark Products Suite Wipes, a high-performance cleaning solution designed for critical applications. These non-sterile, 12 in x 12 in, snag-resistant wipes deliver outstanding absorbance, effectively removing over 98% of surface contamination while minimizing cross-contamination potential to less than 0.02%. Crafted from advanced synthetic Ultra-Fibers, they ensure stringent cleanliness standards are upheld, making them ideal for scientific and controlled environments.