Learn More
Med Vet International Skin Barrier Wipe SionBiotext Isopropyl Alcohol, Water, SD Ethanol -40, Vinyl Caprolactam/VP/Dimethylaminoethyl Methacrylate Copolymer Individual Packet NonSterile, 423784, 100/Box
Catalog No. 503043191
Encompass_Preferred
Helps protect against skin irritation caused by adhesives Leaves a thin breathable film on skin, helping to reduce friction caused by adhesives The film dries within seconds without leaving residue on skin, making it ready for next skin barrier, bandage or dressing application Barrier coating does not have to be removed from skin when changing skin barriers, bandages or dressings
Catalog No. 50-304-3191 Supplier Med Vet International Supplier No. 1229314BX
Helps protect against skin irritation caused by adhesives Leaves a thin breathable film on skin, helping to reduce friction caused by adhesives The film dries within seconds without leaving residue on skin, making it ready for next skin barrier, bandage or dressing application Barrier coating does not have to be removed from skin when changing skin barriers, bandages or dressings