Learn More
Med Vet International PVP Prep Pad First Aid Only 10% Strength Povidone Iodine Individual Packet Medium NonSterile, 12-015-004, 60/Case
Catalog No. 503044340
Encompass_Preferred
Convenient, single-use form ward off infections Non-irritating and virtually non-stinging Wipes measure 1-1/4"x2-1/2" Saturated with a 10% povidone-iodine solution
Catalog No. 50-304-4340 Supplier Med Vet International Supplier No. 1203020CS
Convenient, single-use form ward off infections Non-irritating and virtually non-stinging Wipes measure 1-1/4"x2-1/2" Saturated with a 10% povidone-iodine solution