Learn More
Med Vet International Skin Barrier Wipe Skin-Prep 75 to 100% Strength Isopropyl Alcohol Individual Packet Sterile, 420400, Each
Catalog No. 503045489
Encompass_Preferred
Creates liquid film that protects skin and reduces friction for more comfortable removal of adhesives Smith and Nephew skin prep wipe helps prepare skin for application of new adhesives Can be easily applied just about anywhere, including rounded joints such as knees and elbows Skin prep barrier wipe improves adhesion of tapes and dressings Easily removes by washing with soap and water or skin cleanser
Catalog No. 50-304-5489 Supplier Med Vet International Supplier No. 44723EA
Creates liquid film that protects skin and reduces friction for more comfortable removal of adhesives Smith and Nephew skin prep wipe helps prepare skin for application of new adhesives Can be easily applied just about anywhere, including rounded joints such as knees and elbows Skin prep barrier wipe improves adhesion of tapes and dressings Easily removes by washing with soap and water or skin cleanser