Learn More
Andwin Scientific WIPE SPEC 7 9X9IN PK100
SDP
Catalog No. NC0621605
Encompass_Preferred
The 9x9 in. Spec-Wipe 7 Wipers are excellent for cleaning rough, abrasive, irregular yet delicate surfaces. Its interlock knit design allows durability for vigorous cleaning while its sealed edges reduce lint and fiber.
This item is not returnable. View return policy
Catalog No. NC0621605 Supplier Andwin Scientific Supplier No. 21913214
The 9x9 in. Spec-Wipe 7 Wipers are excellent for cleaning rough, abrasive, irregular yet delicate surfaces. Its interlock knit design allows durability for vigorous cleaning while its sealed edges reduce lint and fiber.