Learn More
Benchmark Products Inc Sterile Suite Wipes, Validated Sterile 10-6, 12 in x 12 in
SDP
Catalog No. NC0165925
Encompass_Preferred
Benchmark Products Inc. sterile Suite wipes offer a unique blend of high-density synthetic Ultra-Fibers, ensuring effective surface cleaning in critical environments. With a validated sterile level, these 12 in x 12 in white wipes remove over 98% of contaminants, minimizing cross-contamination risks. Packed 25 per double bag with 8 bags per case, they stand out for their unrivaled absorbance capacity and non-particulating material, making them ideal for ISO Class 5 use.
This item is not returnable. View return policy
Catalog No. NC0165925 Supplier Benchmark Products Inc Supplier No. WTTC121225CS
Benchmark Products Inc. sterile Suite wipes offer a unique blend of high-density synthetic Ultra-Fibers, ensuring effective surface cleaning in critical environments. With a validated sterile level, these 12 in x 12 in white wipes remove over 98% of contaminants, minimizing cross-contamination risks. Packed 25 per double bag with 8 bags per case, they stand out for their unrivaled absorbance capacity and non-particulating material, making them ideal for ISO Class 5 use.