Learn More
Essentra Porous Technologies Polyester Cellulose Wipes (C1), 300/bag, 12bags/case
SDP
Catalog No. NC0668073
Encompass_Preferred
Polyester Cellulose Wipes (C1) | Nonwoven, hydroentangled | 55% cellulose / 45% polyester | Very low extractables and ion levels | Free of binders and additives
This item is not returnable. View return policy
Catalog No. NC0668073 Supplier Essentra Porous Technologies Supplier No. 7C3099L00
Polyester Cellulose Wipes (C1) | Nonwoven, hydroentangled | 55% cellulose / 45% polyester | Very low extractables and ion levels | Free of binders and additives