Learn More
TEKNIPURE LLC WIPER, TEKNIKPURE, POLYESTER/C
Catalog No. NC2086843
Encompass_Preferred
Wiper, Teknikpure, Polyester/Cellulose Blend Nonwoven Cleanroom Wiper, Size: 12"x12", 3000/cs
This item is not returnable. View return policy
Catalog No. NC2086843 Supplier TEKNIPURE LLC Supplier No. TZ1PCS112
Wiper, Teknikpure, Polyester/Cellulose Blend Nonwoven Cleanroom Wiper, Size: 12"x12", 3000/cs