Learn More
Andwin Scientific WIPERS SPEC 4 STER 12X12 CS300
SDP
Catalog No. NC0092750
Encompass_Preferred
Spec-Wipe 4 Sterile Wipers Ideal for aseptic environments. Soft durable tightly spun continuous-filament 100% double-knit polyester. Clean-cut edges. ISO Class 4 cleanroom-laundered. Sterilized by gamma radiation to SAL 1e-6 in accordance with ANSI/IS
This item is not returnable. View return policy
Catalog No. NC0092750 Supplier Andwin Scientific Supplier No. 21914772
Spec-Wipe 4 Sterile Wipers Ideal for aseptic environments. Soft durable tightly spun continuous-filament 100% double-knit polyester. Clean-cut edges. ISO Class 4 cleanroom-laundered. Sterilized by gamma radiation to SAL 1e-6 in accordance with ANSI/IS