Learn More
Med Vet International Sani-Hands Hand Sanitizing Wipes, 20 Count, Ethyl Alcohol, Soft Pack, P71520, 48/Case
Catalog No. 503043307
Encompass_Preferred
Sani-Hands hand sanitizing wipes clean hands thoroughly, removing soil as well as germs and bacteria A 70 percent ethyl alcohol solution offers effective germ-killing power The solution dries rapidly, making wipes safe to use before eating or drinking Aloe and vitamin E soothe the skin and restore moisture Ethyl alcohol hand wipes are individually wrapped, preventing drying out and providing convenient dispensing and portability These 5.5 x 8.4-inch wipes meet OSHA standards for bloodborne pathogens, making them suitable for professional clinics A soft pack contains 20 wipes
Catalog No. 50-304-3307 Supplier Med Vet International Supplier No. 812687CS
Sani-Hands hand sanitizing wipes clean hands thoroughly, removing soil as well as germs and bacteria A 70 percent ethyl alcohol solution offers effective germ-killing power The solution dries rapidly, making wipes safe to use before eating or drinking Aloe and vitamin E soothe the skin and restore moisture Ethyl alcohol hand wipes are individually wrapped, preventing drying out and providing convenient dispensing and portability These 5.5 x 8.4-inch wipes meet OSHA standards for bloodborne pathogens, making them suitable for professional clinics A soft pack contains 20 wipes