Learn More
ACL Inc WIPES IPA 6/CS
Catalog No. 501037883
Encompass_Preferred
Ideal for removing solder pastes, inks, adhesives and flux residues. Removes oil, grease, ionic and non-ionic residues. Safe for Plastics & Metals. Suitable for cleaning Printed Circuit Boards and hardline coaxial cables as well as restoring automatic and semi-automatic printer stencils to a spotless state. No cotton: 50/50 of polyester and cellulose for low-linting.
This item is not returnable. View return policy
Catalog No. 50-103-7883 Supplier ACL Inc Supplier No. 7600
Ideal for removing solder pastes, inks, adhesives and flux residues. Removes oil, grease, ionic and non-ionic residues. Safe for Plastics & Metals. Suitable for cleaning Printed Circuit Boards and hardline coaxial cables as well as restoring automatic and semi-automatic printer stencils to a spotless state. No cotton: 50/50 of polyester and cellulose for low-linting.