Learn More
Med Vet International Rescue 6" x 7" Wipes, One Step Disinfectant Cleaner and Deodorizer, 160 Count
Supplier:  Med Vet International PHVRESCUEWIPE
Encompass_Preferred
Rescue Wipes powered by Accelerated Hydrogen Peroxide (AHP) provides an easy, convenient and effective way to disinfect your surfaces. Ideal for use on small surfaces like exam tables, stethoscopes, and cage spot cleaning. Quick 1 minute contact time for most pathogens Safe to use around staff and animals Compatible with a wide range of materials  
Catalog No. 50-304-3022