Learn More
MSC MicroCare Stencil Cleaning Wipe, 12 x 17 inch, Polyester/Cellulose, 50 Count
Catalog No. 502836868
Encompass
This stencil cleaning wipe is designed for efficient cleaning of SMT stencils. Made from a durable polyester/cellulose blend, these extra-large wipes measure 12 x 17 inches (305 x 432 mm) and come in packs of 50. Their textured surface provides superior scrubbing ability for fast and effective cleaning in electronics assembly and other precision applications.
  • Large 12 x 17 inch (305 x 432 mm) size.
  • Made from polyester/cellulose for durability and absorbency.
  • Textured surface for enhanced scrubbing.
  • Ideal for SMT stencil cleaning.
  • Supplied as a pack of 50 wipes.
  • Lint-free.
  • Non-woven construction.
  • Cut, not sealed edges.
Catalog No. 50-283-6868 Supplier MSC Supplier No. 57099772
This stencil cleaning wipe is designed for efficient cleaning of SMT stencils. Made from a durable polyester/cellulose blend, these extra-large wipes measure 12 x 17 inches (305 x 432 mm) and come in packs of 50. Their textured surface provides superior scrubbing ability for fast and effective cleaning in electronics assembly and other precision applications.
  • Large 12 x 17 inch (305 x 432 mm) size.
  • Made from polyester/cellulose for durability and absorbency.
  • Textured surface for enhanced scrubbing.
  • Ideal for SMT stencil cleaning.
  • Supplied as a pack of 50 wipes.
  • Lint-free.
  • Non-woven construction.
  • Cut, not sealed edges.