Learn More
Med Vet International Clorox Germicidal Wipes 6 Canisters/Case
Supplier:  Med Vet International CLO35309 / NEW
Encompass_Preferred
Clorox hospital- grade wipes with a 0.55% sodium hypochlorite disinfectant.Packed in a pop-up tub, the wipes are heavy and strong.Tested for compliance with safety, efficacy, shelf life and label strength standards.Kills C. difficile spore within 5 minutes.**Based on Federal EPA and State Registrations as of June 2011. * C. difficile spore claim has been registered by the Federal EPA and may not yet be available in all 50 states. Please check with your sales representative for updates in your state. Use as directed on pre-cleaned, hard, non-porous surfaces. 70 Wipes/Canister, 6 Canisters/Case
Catalog No. NC2746065