Learn More
Essendant Oxivir TB Disinfectant Wipes Refill, 11 x 12, Unscented, White, 160 Wipes/Refill Pouch, 4 Refill Pouches/Carton
Catalog No. 2790587
Encompass_Preferred
Ready-to-use disinfectant cleaner wipe based on proprietary hydrogen peroxide (AHP) technology to deliver fast, effective cleaning performance. Disinfects in 60 seconds. Virucide, bactericide, tuberculocide, fungicide and non-food contact sanitizer. Kills MRSA and Norovirus. Meets bloodborne pathogen standards for decontaminating blood and body fluids. No VOCs and no fragrance added. Non-irritating to skin and eyes.
Catalog No. 27-905-87 Supplier Essendant Supplier No. DVO100823906
Ready-to-use disinfectant cleaner wipe based on proprietary hydrogen peroxide (AHP) technology to deliver fast, effective cleaning performance. Disinfects in 60 seconds. Virucide, bactericide, tuberculocide, fungicide and non-food contact sanitizer. Kills MRSA and Norovirus. Meets bloodborne pathogen standards for decontaminating blood and body fluids. No VOCs and no fragrance added. Non-irritating to skin and eyes.