Learn More
Essendant WIPESKIM ALL PURP BRGBL
Supplier:  Essendant KCC33352
Encompass_Preferred
If you have a task where wiping precision and reliability are important WypAll Oil Grease and Ink Cloths are an excellent choice They are designed for wiping tasks where low lint or the use of chemicals is required as the polypropylene sheets are specially engineered for critical applications involving solvents inks and paints These absorbent sheets are acid- base- and solvent-resistant and ideal for use with lubricants oil and grease Whether you re responsible for cleaning by hand manufacturing/assembly pre-packaging wipe-down or refinishing these industrial wipers are essential tools of the trade BRAG Box Towel/Wipe Type- Dry Wipes Applicable Material- Multiple Material(s)- Polypropylene Number of Plies- 1
Catalog No. 50-000-33912
This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.