Learn More
Essendant W4 Critical Task Wipers, Flat Double Bag, 12 x 12, Unscented, White, 100/Bag, 5 Bags/Carton
Catalog No. 2679953
Encompass_Preferred
Do you require a wiper for a sensitive environment, like cleanrooms, medical equipment manufacturing or the biotech industry? Kimtech Pure W4 Critical Task Dry Wipers could be right for your business. They are specially designed for ISO Class 4 and higher cleanrooms, since they are anti-stat, double bagged for extra protection and low lint (good for environments where ¿lint-free" is important). Their unique, textured surface is absorbent, so you can use them for a variety of tasks. These disposable wipers are ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries that require critical tasks.
Catalog No. 26-799-53 Supplier Essendant Supplier No. KCC33330
Do you require a wiper for a sensitive environment, like cleanrooms, medical equipment manufacturing or the biotech industry? Kimtech Pure W4 Critical Task Dry Wipers could be right for your business. They are specially designed for ISO Class 4 and higher cleanrooms, since they are anti-stat, double bagged for extra protection and low lint (good for environments where ¿lint-free" is important). Their unique, textured surface is absorbent, so you can use them for a variety of tasks. These disposable wipers are ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries that require critical tasks.