Learn More
Essendant WIPESVERAQTFLD12X13WE
Catalog No. 5000021452
Encompass_Preferred
Perfectly suited for day-to-day use in any industry Provide a clean residue free wiper every time with a proven track record of quality and consistency Easily remove any kind of fluid or spill without the need to use several wipes to complete the task Extremely strong both wet and dry and won t fall apart ensuring that the surface cleaned is free of any residues and contaminants Low-linting and specially designed for the areas where quality is critical Smooth Texture 1/4 fold Towel/Wipe Type- Folded Wipers Fold Types- Quarter-Fold Material(s)- Hydro-Entangled Wood Pulp/Polyester Number of Plies- 1
Catalog No. 50-000-21452 Supplier Essendant Supplier No. CHI8710
Perfectly suited for day-to-day use in any industry Provide a clean residue free wiper every time with a proven track record of quality and consistency Easily remove any kind of fluid or spill without the need to use several wipes to complete the task Extremely strong both wet and dry and won t fall apart ensuring that the surface cleaned is free of any residues and contaminants Low-linting and specially designed for the areas where quality is critical Smooth Texture 1/4 fold Towel/Wipe Type- Folded Wipers Fold Types- Quarter-Fold Material(s)- Hydro-Entangled Wood Pulp/Polyester Number of Plies- 1