Learn More
Aviva Systems Biology Corp WIPI1 Antibody - N-terminal region (ARP57075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WIPI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WIPI1. Alternate names: ATG18, FLJ10055, WIPI49, ATG18AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62073