Learn More
Chemglass Life Sciences Wire, 5ft Lgth, Bonding and Grounding, Carbon Steel, Clear Vinyl Insulation, with 5/8" Copper Alligator Clip on each end
SDP
Catalog No. 501531366
Encompass_Preferred
Wire, 5ft Lgth, Bonding and Grounding, Carbon Steel, Clear Vinyl Insulation, with 5/8" Copper Alligator Clip on each end. Carbon steel bonding and grounding wire, 3/16" thick (with the clear, vinyl insulation), and 5/8" copper alligator clamp on each end.
Catalog No. 50-153-1366 Supplier Chemglass Life Sciences Supplier No. CG1978P985
Wire, 5ft Lgth, Bonding and Grounding, Carbon Steel, Clear Vinyl Insulation, with 5/8" Copper Alligator Clip on each end. Carbon steel bonding and grounding wire, 3/16" thick (with the clear, vinyl insulation), and 5/8" copper alligator clamp on each end.