Learn More
Electron Microscopy Sciences Wire Bending Pliers
SDP
Catalog No. 5030558
Encompass
These extra heavy-duty pliers are used for bending any metal wire at a V' shaped angle, allowing for the formation of filaments in accordance to your specification.
Catalog No. 50-305-58 Supplier Electron Microscopy Sciences Supplier No. 62185
These extra heavy-duty pliers are used for bending any metal wire at a V' shaped angle, allowing for the formation of filaments in accordance to your specification.