Description
Includes
ceramic, Bunsen, distribution, heat, chemistry
- Wire gauze is used with Bunsen burners to aid in the distribution of heat
- The ceramic center helps prevent flame from coming into direct contact with the container being heated
- Helps protect lab equipment
- Features rolled smooth wire edges and clean white center ceramic
Specifications
Specifications
| Product Type | Wire Gauze |
| For Use With (Application) | Distribution of Heat with Bunsen Burners |
| Material | Ceramic |
| Length (English) | 5 in. |
| Width (English) | 5 in. |
| Description | Rolled smooth wire edges |
| Dimensions (L x W) | 5 x 5 in. |