Learn More
Research Products International Corp Wire Drying Rack
SDP
Catalog No. 501537765
Encompass
Self standing wire rack is ideal for draining tubes, beakers, flasks and other containers. Prevents glassware from chipping, cracking or breaking. Holds up to 90 items on vinyl coated steel hooks. Place self-standing rack on a table, near a sink or use hooks to hang rack on the wall. Durable rack features heavy duty construction with polystyrene tray and back. Spill pan cleans easily and is removable.
Catalog No. 50-153-7765 Supplier Research Products International Corp Supplier No. 212622
Self standing wire rack is ideal for draining tubes, beakers, flasks and other containers. Prevents glassware from chipping, cracking or breaking. Holds up to 90 items on vinyl coated steel hooks. Place self-standing rack on a table, near a sink or use hooks to hang rack on the wall. Durable rack features heavy duty construction with polystyrene tray and back. Spill pan cleans easily and is removable.