Learn More
Electron Microscopy Sciences Winlab Slide Rack Wire Handles for 20 Slide Rack
SDP
Catalog No. 50931060
Encompass_Preferred
Steel wire is bent to form a handle, use these handles for manual staining and the Medite stainer, Model W3-6 fits all WINLAB 20-Slide Racks.
Capacity: 20 slides
Catalog No. 50-931-060 Supplier Electron Microscopy Sciences Supplier No. 7143720
Add to Cart
Add to Cart
Steel wire is bent to form a handle, use these handles for manual staining and the Medite stainer, Model W3-6 fits all WINLAB 20-Slide Racks.
Capacity: 20 slides