Learn More
Electron Microscopy Sciences Winlab Slide Rack Wire Handles for 30 Slide Rack
SDP
Catalog No. 50931061
Encompass_Preferred
Steel wire is bent to form a handle, use these handles for manual staining and the Medite stainer, Model W3-5 fits all WINLAB 30-Slide Racks.
Capacity: 30 slides
Catalog No. 50-931-061 Supplier Electron Microscopy Sciences Supplier No. 7143730
Steel wire is bent to form a handle, use these handles for manual staining and the Medite stainer, Model W3-5 fits all WINLAB 30-Slide Racks.
Capacity: 30 slides