Learn More
Electron Microscopy Sciences Wire Looping Pliers
SDP
Catalog No. 5031060
Encompass
Stainless steel wire looping pliers, enables the user to bend wire easily. The round jaws in one and the boxjointed for durability. Jaws are finely polished, one being round and the other concave.
Catalog No. 50-310-60 Supplier Electron Microscopy Sciences Supplier No. 6218610
Stainless steel wire looping pliers, enables the user to bend wire easily. The round jaws in one and the boxjointed for durability. Jaws are finely polished, one being round and the other concave.