Learn More
Grainger Nsi Industries Wire Marker Book, Vinyl Impregnated Cloth, 0-9 Legend, 450/Pk.
Catalog No. 502774441
Encompass_Preferred
This wire marker book is designed for the efficient identification of electrical wiring and components. Each book contains 450 pre-printed markers featuring the digits 0 to 9, allowing for clear and organized labeling. Constructed from vinyl impregnated cloth film, these markers are built for durability in various environments.
  • Features 450 individual wire markers
  • Includes digits 0 to 9 for comprehensive identification
  • Strong adhesive ensures lasting labels
  • Material consists of durable vinyl impregnated cloth film
  • Operating temperature range of -40°F to 250°F
  • Compact pocket pal design for easy transport
  • Made in the United States of America
  • ARRA and Buy American Act compliant
Catalog No. 50-277-4441 Supplier Grainger Supplier No. 174M51
This wire marker book is designed for the efficient identification of electrical wiring and components. Each book contains 450 pre-printed markers featuring the digits 0 to 9, allowing for clear and organized labeling. Constructed from vinyl impregnated cloth film, these markers are built for durability in various environments.
  • Features 450 individual wire markers
  • Includes digits 0 to 9 for comprehensive identification
  • Strong adhesive ensures lasting labels
  • Material consists of durable vinyl impregnated cloth film
  • Operating temperature range of -40°F to 250°F
  • Compact pocket pal design for easy transport
  • Made in the United States of America
  • ARRA and Buy American Act compliant