Learn More
Grainger Nsi Industries Wire Marker Book, Vinyl Impregnated Cloth, 450 Markers, 10 Pages
Catalog No. 502777110
Encompass_Preferred
This wire marker book contains 450 preprinted markers designed for effective electrical identification and organization. The markers are manufactured from durable vinyl impregnated cloth and feature a black legend on a white background for high visibility.
  • Includes 10 pages with a total of 450 individual markers
  • Legends include 0 to 9, L1, L2, L3, T1, T2, and T3
  • Features strong adhesive for secure placement
  • Operates in temperatures from -40°F to 250°F
  • Made in the USA
Catalog No. 50-277-7110 Supplier Grainger Supplier No. 174M54
This wire marker book contains 450 preprinted markers designed for effective electrical identification and organization. The markers are manufactured from durable vinyl impregnated cloth and feature a black legend on a white background for high visibility.
  • Includes 10 pages with a total of 450 individual markers
  • Legends include 0 to 9, L1, L2, L3, T1, T2, and T3
  • Features strong adhesive for secure placement
  • Operates in temperatures from -40°F to 250°F
  • Made in the USA