Learn More
Sklar Instruments Wire Net Basket, Sklar, 13 in. x 10 in. x 2-3/4 in. Wire Net Basket 340mm x 250mm x 70mm, 6.5mm Grid Openings, Stackable. Sklar's Wire Net Basket is designed for use with cleaning, sterilizing, and handling small instruments. EA/1.
SDP
Catalog No. 501437270 Shop All Sklar Instruments Products
Encompass_Preferred
Wire Net Basket, Sklar, 13 in. x 10 in. x 2-3/4 in. Wire Net Basket 340mm x 250mm x 70mm, 6.5mm Grid Openings, Stackable. Sklar's Wire Net Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, and lids are sold separately.
This item is not returnable. View return policy
Catalog No. 50-143-7270 Supplier Sklar Instruments Supplier No. 106672
Wire Net Basket, Sklar, 13 in. x 10 in. x 2-3/4 in. Wire Net Basket 340mm x 250mm x 70mm, 6.5mm Grid Openings, Stackable. Sklar's Wire Net Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, and lids are sold separately.