Learn More
Ace Glass, Inc. Wire probe, K thermocouple, 120in, fiberglass covered, male miniplug
SDP
Catalog No. 50983326
Encompass
120 insulated wire temperature probe with type K thermocouple is fiberglass covered and has a male mini plug. Provides measurement from -20 degrees C to 550 degrees C, with more corrosion resistance than type J probes.
Catalog No. 50-983-326 Supplier Ace Glass, Inc. Supplier No. 1211329
Add to Cart
Add to Cart
120 insulated wire temperature probe with type K thermocouple is fiberglass covered and has a male mini plug. Provides measurement from -20 degrees C to 550 degrees C, with more corrosion resistance than type J probes.