Learn More
Ace Glass, Inc. Wire probe, T thermocouple, 120in, PTFE covered, male mini plug
SDP
Catalog No. 50977464
Encompass
120 insulated wire temperature probe with type T thermocouple is PTFE-covered and has a male mini plug. Provides measurement from -200 degrees C to 200 degrees C.
Catalog No. 50-977-464 Supplier Ace Glass, Inc. Supplier No. 1211382
120 insulated wire temperature probe with type T thermocouple is PTFE-covered and has a male mini plug. Provides measurement from -200 degrees C to 200 degrees C.