Learn More
MSC Value Wire Rope Clip, 1/8 in, Stainless Steel, 5 Pack
Catalog No. 502869470
Encompass
This wire rope clip is designed to securely fasten the loose end of a loop of wire rope. Constructed from durable stainless steel, it offers excellent corrosion resistance and strength, making it suitable for various demanding applications. This clip is specifically engineered for use with 1/8 in diameter wire rope.
  • Securely fastens wire rope ends
  • Constructed from durable stainless steel
  • Designed for 1/8 in diameter wire rope
  • Provides strong and reliable connection
Catalog No. 50-286-9470 Supplier MSC Supplier No. 53589396
This wire rope clip is designed to securely fasten the loose end of a loop of wire rope. Constructed from durable stainless steel, it offers excellent corrosion resistance and strength, making it suitable for various demanding applications. This clip is specifically engineered for use with 1/8 in diameter wire rope.
  • Securely fastens wire rope ends
  • Constructed from durable stainless steel
  • Designed for 1/8 in diameter wire rope
  • Provides strong and reliable connection