Learn More
MSC Value Collection Wire Rope Clip, 7/8 in. Rope Diameter, Steel, Galvanized, 1 ea.
Catalog No. 5000511396
Encompass
This wire rope clip is designed to securely fasten 7/8 in. diameter rope. Constructed from durable steel with a galvanized finish, it provides robust performance for various rigging and fastening applications.
  • Compatible with 7/8 in. diameter rope
  • Durable steel construction
  • Galvanized finish for corrosion resistance
  • Reliable holding strength for wire rope applications
Catalog No. 50-005-11396 Supplier MSC Supplier No. 53593216
This wire rope clip is designed to securely fasten 7/8 in. diameter rope. Constructed from durable steel with a galvanized finish, it provides robust performance for various rigging and fastening applications.
  • Compatible with 7/8 in. diameter rope
  • Durable steel construction
  • Galvanized finish for corrosion resistance
  • Reliable holding strength for wire rope applications