Learn More
Grainger Wire Rope Locking Stud LH Thread 1/16 in
Catalog No. 502767772
Encompass_Preferred
The Loos & Co. Wire Rope Thimble is made of galvanized steel with a 3/32 inch inside diameter. It is designed to fit 3/32 inch wire rope and protect it from wear. The thimble has a closed eye and is used to form eyes on wire rope for use with shackles, hooks, and other hardware. It provides a hard wearing surface to prevent crushing and deformation of the wire rope. This wire rope thimble is galvanized for corrosion resistance. It meets military specification MIL-T-781J and is made in the USA by Loos & Co., a leading manufacturer of wire rope fittings.
Catalog No. 50-276-7772 Supplier Grainger Supplier No. 16X710
The Loos & Co. Wire Rope Thimble is made of galvanized steel with a 3/32 inch inside diameter. It is designed to fit 3/32 inch wire rope and protect it from wear. The thimble has a closed eye and is used to form eyes on wire rope for use with shackles, hooks, and other hardware. It provides a hard wearing surface to prevent crushing and deformation of the wire rope. This wire rope thimble is galvanized for corrosion resistance. It meets military specification MIL-T-781J and is made in the USA by Loos & Co., a leading manufacturer of wire rope fittings.