Learn More
Grainger Wire Rope Locking Stud LH Thread 3/32 in
Catalog No. 502768255
Encompass_Preferred
The Loos & Co. Wire Rope Thimble is made of galvanized steel with a 3/32" inside diameter. It is designed to fit 3/32" wire rope and protect it from wear. The thimble has a closed eye and is used to form eyes on wire rope for use with shackles, hooks, and other hardware. It helps prevent kinking and damage to the wire rope. This galvanized steel thimble is very durable for heavy load applications. The dimensions are 1-1/4" high x 1-3/8" wide x 1/2" thick. It weighs approximately 0.4 lbs. This is an industrial grade thimble made in the USA by Loos & Co., a leading manufacturer of rigging products.
Catalog No. 50-276-8255 Supplier Grainger Supplier No. 16X714
The Loos & Co. Wire Rope Thimble is made of galvanized steel with a 3/32" inside diameter. It is designed to fit 3/32" wire rope and protect it from wear. The thimble has a closed eye and is used to form eyes on wire rope for use with shackles, hooks, and other hardware. It helps prevent kinking and damage to the wire rope. This galvanized steel thimble is very durable for heavy load applications. The dimensions are 1-1/4" high x 1-3/8" wide x 1/2" thick. It weighs approximately 0.4 lbs. This is an industrial grade thimble made in the USA by Loos & Co., a leading manufacturer of rigging products.