Learn More
Grainger Wire Rope Locking Stud LH Thread 3/8 in
Catalog No. 502768256
Encompass_Preferred
The Grainger 16X738 is a 16-inch diameter, 7/8-inch arbor hole, 38 grit aluminum oxide grinding wheel. It is rated for a maximum safe speed of 6,600 RPM. This thin wheel is 1/4 inch thick and is designed for general purpose grinding operations on ferrous metals. The aluminum oxide abrasive cuts quickly with moderate wheel life. This is a type 27 depressed center grinding wheel suitable for bench grinders, pedestal grinders, and other stationary grinding machines.
Catalog No. 50-276-8256 Supplier Grainger Supplier No. 16X738
The Grainger 16X738 is a 16-inch diameter, 7/8-inch arbor hole, 38 grit aluminum oxide grinding wheel. It is rated for a maximum safe speed of 6,600 RPM. This thin wheel is 1/4 inch thick and is designed for general purpose grinding operations on ferrous metals. The aluminum oxide abrasive cuts quickly with moderate wheel life. This is a type 27 depressed center grinding wheel suitable for bench grinders, pedestal grinders, and other stationary grinding machines.