Learn More
Grainger Wire Rope Locking Stud LH Thread 1/16 in
Catalog No. 502765684
Encompass_Preferred
The Grainger Approved Wire Rope Locking Stud is a 16mm wire rope stud made of galvanized steel with a working load limit of 1,770 lbs. It features a threaded stud design that allows for adjustable positioning on wire rope and aircraft cable. This wire rope stud can be used for rigging and securing applications. It is zinc plated for corrosion resistance.
Catalog No. 50-276-5684 Supplier Grainger Supplier No. 16X712
The Grainger Approved Wire Rope Locking Stud is a 16mm wire rope stud made of galvanized steel with a working load limit of 1,770 lbs. It features a threaded stud design that allows for adjustable positioning on wire rope and aircraft cable. This wire rope stud can be used for rigging and securing applications. It is zinc plated for corrosion resistance.