Learn More
Grainger Wire Rope Locking Stud LH Thread 1/8 in
Catalog No. 502768198
Encompass_Preferred
The Grainger 1/8" Swage Stud Fittings for Wire Rope are high-strength fittings designed to terminate wire rope for lifting and rigging applications. They feature a forged carbon steel construction and are zinc plated for corrosion resistance. The fittings have a working load limit up to 1,650 lbs and are available in a variety of styles including eye, fork, and hook configurations. The swage stud design allows for secure attachment to wire rope without splicing. Overall, these fittings provide a durable and reliable option for creating terminations and connections with 1/8" diameter wire rope.
Catalog No. 50-276-8198 Supplier Grainger Supplier No. 16X720
The Grainger 1/8" Swage Stud Fittings for Wire Rope are high-strength fittings designed to terminate wire rope for lifting and rigging applications. They feature a forged carbon steel construction and are zinc plated for corrosion resistance. The fittings have a working load limit up to 1,650 lbs and are available in a variety of styles including eye, fork, and hook configurations. The swage stud design allows for secure attachment to wire rope without splicing. Overall, these fittings provide a durable and reliable option for creating terminations and connections with 1/8" diameter wire rope.