Learn More
Grainger Wire Rope Locking Stud LH Thread 5/32 in
Catalog No. 502765659
Encompass_Preferred
The Grainger 3/16" swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged end that grips the wire rope for a secure connection. The stud end has coarse threads for attaching other hardware. These zinc plated fittings meet military specs and are available in a variety of sizes with breaking strengths from 1,740 lbs to 9,800 lbs. They provide a reliable, heavy duty end fitting for 3/16" wire rope in lifting, pulling, and positioning applications.
Catalog No. 50-276-5659 Supplier Grainger Supplier No. 16X724
The Grainger 3/16" swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged end that grips the wire rope for a secure connection. The stud end has coarse threads for attaching other hardware. These zinc plated fittings meet military specs and are available in a variety of sizes with breaking strengths from 1,740 lbs to 9,800 lbs. They provide a reliable, heavy duty end fitting for 3/16" wire rope in lifting, pulling, and positioning applications.