Learn More
Grainger Wire Rope Locking Stud LH Thread 1/8 in
Catalog No. 502765118
Encompass_Preferred
The product is a Crosby 9/16"-18 Wire Rope Clip, made of galvanized steel. It is used to attach wire rope to various fittings. The clip has a working load limit of 3,150 lbs and is rated for 6x19 and 6x36 class wire rope. The clip measures 2-1/4" long x 1-3/16" wide x 15/16" high. It features a heavy malleable iron saddle with U-bolt and nuts. The clip conforms to Fed Spec RR-C-271 Type I Class A. It is manufactured in the USA by The Crosby Group LLC.
Catalog No. 50-276-5118 Supplier Grainger Supplier No. 16X718
The product is a Crosby 9/16"-18 Wire Rope Clip, made of galvanized steel. It is used to attach wire rope to various fittings. The clip has a working load limit of 3,150 lbs and is rated for 6x19 and 6x36 class wire rope. The clip measures 2-1/4" long x 1-3/16" wide x 15/16" high. It features a heavy malleable iron saddle with U-bolt and nuts. The clip conforms to Fed Spec RR-C-271 Type I Class A. It is manufactured in the USA by The Crosby Group LLC.